Assay Details
Target Gene Details
Entrez Gene ID: | 66628 |
Gene Name: | tRNA-histidine guanylyltransferase 1-like (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.11:45945305-45955503 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thg1l | NM_001080969.3 | 3 | 490 | NP_001074438.1 |
| NM_001290737.1 | 4 | 433 | NP_001277666.1 | |
| AK007233.1 | 5 | 471 | BAB24907.1 | |
| AK010876.1 | 3 | 482 | BAB27240.1 | |
| AK015467.1 | 3 | 284 | BAB29859.1 | |
| AK016076.1 | 4 | 434 | BAB30104.1 | |
| AK017532.1 | 3 | 473 | BAC25519.1 | |
| BC092541.1 | 3 | 470 | AAH92541.1 | |
| BC115560.1 | 3 | 478 | ||
| BC115561.1 | 3 | 478 |

Genomic Map