Assay Details
Target Gene Details
Entrez Gene ID: | 75909 |
Gene Name: | vacuole membrane protein 1 |
Gene Aliases: |
3110098I04Rik, 4930579A11Rik, AI787464, Tango5, Tmem49, mir-21a, ni-2 |
Location: |
Chr.11:86583852-86683822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vmp1 | NM_029478.3 | NP_083754.2 | ||
| AB047550.1 | ||||
| AB076609.1 | BAD06453.1 | |||
| AK016312.1 | 9 | 1059 | ||
| AK043091.1 | BAC31456.1 | |||
| AK049559.1 | ||||
| AK077443.1 | BAC36803.1 | |||
| AK159468.1 | BAE35109.1 | |||
| AK167173.1 | ||||
| AK167202.1 | ||||
| AK211254.1 | ||||
| AK218778.1 | ||||
| AK219288.1 | ||||
| AX775963.1 | CAE11681.1 | |||
| BC004013.1 | AAH04013.1 |
More Information
Set Membership: |
|

Genomic Map