Assay Details
Target Gene Details
Entrez Gene ID: | 28084 |
Gene Name: | vacuolar protein sorting 25 (yeast) |
Gene Aliases: |
1110020N13Rik, AW107467, D11Wsu68e |
Location: |
Chr.11:101253611-101259547 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps25 | NM_001284411.1 | NP_001271340.1 | ||
| NM_001284412.1 | NP_001271341.1 | |||
| NM_001284414.1 | NP_001271343.1 | |||
| NM_026776.4 | NP_081052.2 | |||
| AK002500.1 | BAB22148.1 | |||
| AK003715.2 | BAB22955.1 | |||
| AK003877.1 | BAB23054.1 | |||
| AK005149.1 | ||||
| AK082933.1 | ||||
| AK145811.1 | BAE26665.1 | |||
| AK150822.1 | BAE29884.1 | |||
| AK162230.1 | ||||
| AK166008.1 | BAE38516.1 | |||
| AK190530.1 | ||||
| AK214794.1 | ||||
| BC029181.1 | AAH29181.1 |
More Information
Set Membership: |
|

Genomic Map