Assay Details
Target Gene Details
Entrez Gene ID: | 67178 |
Gene Name: | zinc finger, matrin type 5 |
Gene Aliases: |
2610510L01Rik, D11Bwg0572e, D11Bwg1548e |
Location: |
Chr.11:4704606-4737669 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmat5 | NM_026015.2 | NP_080291.1 | ||
| AK003361.1 | BAB22736.1 | |||
| AK007565.1 | BAB25111.1 | |||
| AK012098.1 | BAB28029.1 | |||
| AK051823.1 | ||||
| BC031796.1 | AAH31796.1 |

Genomic Map