Assay Details
Target Gene Details
Entrez Gene ID: | 72149 |
Gene Name: | STE20-related kinase adaptor alpha |
Gene Aliases: |
2610019A05Rik, 6030402H20Rik, AI480680, E130112C09Rik |
Location: |
Chr.11:106162974-106193604 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strada | NM_001252448.1 | NP_001239377.1 | ||
| NM_001252449.1 | NP_001239378.1 | |||
| NM_028126.3 | NP_082402.1 | |||
| AK011451.1 | BAB27626.1 | |||
| AK138358.1 | BAE23631.1 | |||
| AY341872.1 | AAQ24157.1 | |||
| AY341878.1 | AAQ24162.1 | |||
| BC058517.1 | AAH58517.1 |
More Information
Set Membership: |
|

Genomic Map