Assay Details
Target Gene Details
Entrez Gene ID: | 76795 |
Gene Name: | TBC1 domain family, member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:50131057-50172785 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d9b | NM_001290759.1 | 3 | 370 | NP_001277688.1 |
| NM_001290760.1 | 3 | 370 | NP_001277689.1 | |
| NM_029745.2 | 3 | 370 | NP_084021.2 | |
| AK035457.1 | 3 | 335 | BAC29068.1 | |
| AK049999.1 | 3 | 330 | BAC34024.2 | |
| AK147964.1 | 3 | 335 | BAE28254.1 | |
| AK166472.1 | 3 | 353 | ||
| AK172999.1 | 4 | 310 | BAD32277.1 | |
| AK182181.1 | 2 | 114 | ||
| AK186143.1 | 2 | 179 | ||
| AK195054.1 | ||||
| AK215503.1 | 2 | 114 | ||
| AK215674.1 | 2 | 114 | ||
| BC062928.1 | 3 | 357 | AAH62928.1 | |
| BC065080.1 | 3 | 349 | AAH65080.1 |
More Information
Set Membership: |
|

Genomic Map