Assay Details
Target Gene Details
Entrez Gene ID: | 70450 |
Gene Name: | unc-13 homolog D (C. elegans) |
Gene Aliases: |
2610108D09Rik, Jinx, Munc13-4, mFLJ00067 |
Location: |
Chr.11:116062095-116078195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc13d | NM_001009573.2 | NP_001009573.2 | ||
| AK220199.1 | BAD90124.1 | |||
| BC141289.1 | ||||
| BG146628.1 | ||||
| CF164660.1 | ||||
| EF127645.1 |

Genomic Map