Assay Details
Target Gene Details
Entrez Gene ID: | 56444 |
Gene Name: | ARP10 actin-related protein 10 |
Gene Aliases: |
Actr11, Arp11 |
Location: |
Chr.12:70937857-70964718 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr10 | NM_019785.2 | 12 | 1026 | NP_062759.2 |
| AF190797.1 | 12 | 993 | AAF05617.1 | |
| AK090043.1 | 12 | 1030 | ||
| AK161642.1 | 12 | 986 | BAE36507.1 | |
| AK210085.1 | 4 | 244 | ||
| BC002227.1 | 12 | 986 | AAH02227.1 |
More Information
Set Membership: |
|

Genomic Map