Assay Details
Target Gene Details
Entrez Gene ID: | 107829 |
Gene Name: | THO complex 5 |
Gene Aliases: |
1700060C24Rik, A430085L24Rik, Fmip, PK1.3, Sgk2 |
Location: |
Chr.11:4895283-4928867 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thoc5 | NM_172438.3 | NP_766026.1 | ||
| AK050734.1 | BAC34398.1 | |||
| AK173078.1 | BAD32356.1 | |||
| AK193058.1 | ||||
| AK212973.1 | ||||
| BC039758.1 | AAH39758.1 |

Genomic Map