Assay Details
Target Gene Details
Entrez Gene ID: | 74855 |
Gene Name: | family with sequence similarity 228, member A |
Gene Aliases: |
4930417G10Rik |
Location: |
Chr.12:4697272-4738468 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam228a | NM_029107.2 | 2 | 235 | NP_083383.1 |
| AK015158.1 | 2 | 107 | BAB29728.1 | |
| AK029492.1 | 2 | 236 | BAC26475.1 | |
| AK133272.1 | 2 | 272 | ||
| BC138846.1 | 2 | 170 | ||
| BC138853.1 | 2 | 78 |

Genomic Map