Assay Details
Target Gene Details
Entrez Gene ID: | 66204 |
Gene Name: | acylphosphatase 1, erythrocyte (common) type |
Gene Aliases: |
1110039O14Rik, AI325944 |
Location: |
Chr.12:85272398-85288196 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acyp1 | NM_025421.2 | NP_079697.1 | ||
| AK004176.1 | BAB23208.1 | |||
| AK007244.1 | BAB24911.1 | |||
| AK027969.1 | BAC25681.1 | |||
| BC061088.1 |
Target Gene Details
Entrez Gene ID: | 72350 |
Gene Name: | zinc finger, C2HC-type containing 1C |
Gene Aliases: |
2810002I04Rik, AV046379, Fam164c |
Location: |
Chr.12:85274388-85299361 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc2hc1c |
More Information
Set Membership: |
|

Genomic Map