Assay Details
Target Gene Details
Entrez Gene ID: | 20446 |
Gene Name: | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2 |
Gene Aliases: |
II, ST6GalNAc, Siat7, Siat7b |
Location: |
Chr.11:116676703-116695231 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6galnac2 | NM_009180.3 | 3 | 371 | NP_033206.2 |
| AK004613.1 | 3 | 372 | BAB23410.1 | |
| X93999.1 | 3 | 335 | CAA63821.1 |

Genomic Map