Assay Details
Target Gene Details
Entrez Gene ID: | 117160 |
Gene Name: | tweety homolog 2 (Drosophila) |
Gene Aliases: |
1110001A03Rik |
Location: |
Chr.11:114675468-114720984 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttyh2 | NM_053273.2 | NP_444503.2 | ||
| AF329682.1 | AAL16785.1 | |||
| AK152172.1 | BAE31004.1 | |||
| AK154835.1 | ||||
| AK155166.1 | BAE33089.1 | |||
| AK156393.1 | BAE33698.1 | |||
| AK168408.1 | BAE40325.1 | |||
| AK204126.1 | ||||
| BC060740.1 | AAH60740.1 | |||
| BY731753.1 |
More Information
Set Membership: |
|

Genomic Map