Assay Details
Target Gene Details
Entrez Gene ID: | 53422 |
Gene Name: | Y box protein 2 |
Gene Aliases: |
Msy2 |
Location: |
Chr.11:69935835-69941604 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybx2 | NM_016875.2 | NP_058571.2 | ||
| AF073954.1 | AAC98673.1 | |||
| AF073955.1 | AAC98674.1 | |||
| AK131732.1 | ||||
| AK131786.1 | ||||
| AK161439.1 | BAE36397.1 | |||
| BC100454.1 | AAI00455.1 | |||
| BC141112.1 | ||||
| DV661016.1 |

Genomic Map