Assay Details
Target Gene Details
Entrez Gene ID: | 68385 |
Gene Name: | TLC domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.11:78176716-78180820 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlcd1 | NM_001291235.1 | NP_001278164.1 | ||
| NM_001291236.1 | NP_001278165.1 | |||
| NM_001291237.1 | NP_001278166.1 | |||
| NM_026708.2 | NP_080984.1 | |||
| AA671983.1 | ||||
| AK002265.1 | ||||
| AK002703.1 | BAB22297.1 | |||
| AK046577.1 | BAE20654.1 | |||
| BC005702.1 | AAH05702.1 | |||
| BC039182.1 | AAH39182.1 | |||
| BG083251.2 |

Genomic Map