Assay Details
Target Gene Details
Entrez Gene ID: | 103768 |
Gene Name: | tubulin, gamma 2 |
Gene Aliases: |
AI504772, Tubgl |
Location: |
Chr.11:101155871-101161787 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubg2 | NM_134028.2 | NP_598789.1 | ||
| AB158481.1 | BAD27265.1 | |||
| BC019652.1 | AAH19652.1 | |||
| BC051439.1 | AAH51439.1 |
More Information
Set Membership: |
|

Genomic Map