Assay Details
Target Gene Details
Entrez Gene ID: | 77864 |
Gene Name: | yippee-like 2 (Drosophila) |
Gene Aliases: |
6430570G24, E130113K08Rik |
Location: |
Chr.11:86936425-86993849 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ypel2 | NM_001005341.3 | 1 | 89 | NP_001005341.1 |
| AK021387.1 | 1 | 35 | ||
| AK089126.2 | ||||
| BY768876.1 | 1 | 95 |

Genomic Map