Assay Details
Target Gene Details
Entrez Gene ID: | 407821 |
Gene Name: | zinc and ring finger 3 |
Gene Aliases: |
Gm1167 |
Location: |
Chr.11:5276329-5444847 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Znrf3 | NM_001080924.2 | NP_001074393.1 | ||
| NM_001290501.1 | NP_001277430.1 | |||
| AK133342.1 | BAE21609.1 | |||
| AK163639.1 | ||||
| BC151080.1 | ||||
| BC151083.1 |

Genomic Map