Assay Details
Target Gene Details
Entrez Gene ID: | 75847 |
Gene Name: | isoprenoid synthase domain containing |
Gene Aliases: |
4930579E17Rik, AV040780 |
Location: |
Chr.12:36380585-36689503 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ispd | NM_001289502.1 | NP_001276431.1 | ||
| NM_001289503.1 | NP_001276432.1 | |||
| NM_178629.6 | NP_848744.2 | |||
| AK016317.1 | ||||
| AK039004.1 | 6 | 1146 | BAC30199.1 | |
| AK045940.1 | BAC32538.1 | |||
| AK051322.1 | ||||
| BC024061.1 | AAH24061.1 | |||
| BC086665.1 | AAH86665.1 | |||
| BC118038.1 | 6 | 1046 |
More Information
Set Membership: |
|

Genomic Map