Assay Details
Target Gene Details
Entrez Gene ID: | 68737 |
Gene Name: | angel homolog 1 (Drosophila) |
Gene Aliases: |
1110030H02Rik, mKIAA0759 |
Location: |
Chr.12:86700502-86734187 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angel1 | NM_144524.2 | NP_653107.2 | ||
| AK143549.1 | ||||
| BC019148.1 | AAH19148.1 |
More Information
Set Membership: |
|

Genomic Map