Assay Details
Target Gene Details
Entrez Gene ID: | 216825 |
Gene Name: | ubiquitin specific peptidase 22 |
Gene Aliases: |
AI427806 |
Location: |
Chr.11:61151781-61175059 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp22 | NM_001004143.4 | NP_001004143.2 | ||
| AK034492.1 | ||||
| AK040187.1 | BAE20573.1 | |||
| AK155071.1 | BAE33026.1 | |||
| AK160992.1 | BAE36137.1 | |||
| AK220368.1 | BAD90248.1 | |||
| BC080737.1 | AAH80737.1 |
More Information
Set Membership: |
|

Genomic Map