Assay Details
Target Gene Details
Entrez Gene ID: | 217473 |
Gene Name: | ankyrin repeat and MYND domain containing 2 |
Gene Aliases: |
AI035571, ZMYND20 |
Location: |
Chr.12:36157124-36197291 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankmy2 | NM_146033.3 | NP_666145.3 | ||
| AK037644.1 | BAC29839.1 | |||
| AK077556.1 | BAC36860.1 | |||
| AK164434.1 | BAE37787.1 | |||
| BC012275.1 | AAH12275.1 | |||
| BC024959.1 | AAH24959.1 |

Genomic Map