Assay Details
Target Gene Details
Entrez Gene ID: | 21393 |
Gene Name: | titin-cap |
Gene Aliases: |
AI503981, AJ223855 |
Location: |
Chr.11:98383811-98384953 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcap | NM_011540.2 | 1 | 68 | NP_035670.2 |
| AJ223854.1 | 1 | 68 | CAA11585.1 | |
| AK010167.1 | 1 | 56 | BAB26743.1 | |
| BC027631.1 | AAH27631.1 | |||
| Y15845.1 | 1 | 58 | CAB38077.1 |

Genomic Map