Assay Details
Target Gene Details
Entrez Gene ID: | 75604 |
Gene Name: | transmembrane 4 superfamily member 5 |
Gene Aliases: |
2010003F10Rik, AI323987 |
Location: |
Chr.11:70505274-70511183 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm4sf5 | NM_029360.3 | 1 | 151 | NP_083636.2 |
| AK008064.1 | 1 | 149 | BAB25438.1 | |
| BB869909.1 | 1 | 152 | ||
| BC010782.1 | 1 | 111 | AAH10782.1 |

Genomic Map