Assay Details
Target Gene Details
Entrez Gene ID: | 11565 |
Gene Name: | adenylosuccinate synthetase like 1 |
Gene Aliases: |
AI528595, Adss, Adss1 |
Location: |
Chr.12:112620047-112641355 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adssl1 | NM_007421.2 | 10 | 1076 | NP_031447.1 |
| AK089520.1 | 10 | 1051 | ||
| AK146907.1 | 10 | 1079 | BAE27520.1 | |
| AK149634.1 | 10 | 1051 | ||
| AK150874.1 | 10 | 1047 | BAE29924.1 | |
| BC039943.1 | 10 | 1107 | AAH39943.1 | |
| M74495.1 | 10 | 1034 | AAA82870.1 |
More Information
Set Membership: |
|

Genomic Map