Assay Details
Target Gene Details
Entrez Gene ID: | 20742 |
Gene Name: | spectrin beta, non-erythrocytic 1 |
Gene Aliases: |
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049 |
Location: |
Chr.11:30099395-30268204 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptbn1 | NM_009260.2 | NP_033286.2 | ||
| NM_175836.2 | NP_787030.2 | |||
| AF016040.1 | AAD01566.1 | |||
| AF017112.1 | AAD01616.1 | |||
| AK157727.1 | BAE34172.1 | |||
| AK220456.1 | BAD90485.1 | |||
| M74773.1 | AAC42040.1 |

Genomic Map