Assay Details
Target Gene Details
Entrez Gene ID: | 75604 |
Gene Name: | transmembrane 4 superfamily member 5 |
Gene Aliases: |
2010003F10Rik, AI323987 |
Location: |
Chr.11:70505274-70511183 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm4sf5 | NM_029360.3 | NP_083636.2 | ||
| AI323987.1 | ||||
| AK008064.1 | BAB25438.1 | |||
| BC010782.1 | AAH10782.1 |

Genomic Map