Assay Details
Target Gene Details
Entrez Gene ID: | 195018 |
Gene Name: | zinc finger, ZZ-type with EF hand domain 1 |
Gene Aliases: |
8430405D05Rik, BC051585, C130099L13Rik, C85424, mKIAA0399 |
Location: |
Chr.11:72796226-72927120 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zzef1 | NM_001045536.2 | 3 | 666 | NP_001039001.1 |
| AK158924.1 | 4 | 586 |

Genomic Map