Assay Details
Target Gene Details
Entrez Gene ID: | 217031 |
Gene Name: | transcriptional adaptor 2A |
Gene Aliases: |
AV319371, D030022J10Rik, Tada2l |
Location: |
Chr.11:84078920-84129642 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tada2a | NM_172562.3 | NP_766150.1 | ||
| AK157938.1 | BAE34272.1 | |||
| AK216505.1 | ||||
| BC025448.1 | AAH25448.1 | |||
| BC038821.1 | AAH38821.1 |
More Information
Set Membership: |
|

Genomic Map