Assay Details
Target Gene Details
Entrez Gene ID: | 217353 |
Gene Name: | transmembrane channel-like gene family 6 |
Gene Aliases: |
D11Ertd204e, EVER1 |
Location: |
Chr.11:117765985-117782193 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc6 | NM_145439.2 | NP_663414.3 | ||
| NM_181321.3 | NP_851838.1 | |||
| AK052814.1 | BAC35157.1 | |||
| AK077671.1 | BAC36945.1 | |||
| AK090179.1 | ||||
| AY236497.1 | AAP69875.1 | |||
| AY263158.1 | AAP78773.1 | |||
| BC004840.1 | AAH04840.2 | |||
| BC058195.1 | AAH58195.1 | |||
| BY345160.1 |

Genomic Map