Assay Details
Target Gene Details
Entrez Gene ID: | 319939 |
Gene Name: | tensin 3 |
Gene Aliases: |
BC023928, F830010I22Rik, TEM6, Tens1 |
Location: |
Chr.11:8431652-8664735 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tns3 | NM_001083587.1 | NP_001077056.1 | ||
| AK158101.1 | BAE34356.1 |
More Information
Set Membership: |
|

Genomic Map