Assay Details
Target Gene Details
Entrez Gene ID: | 20850 |
Gene Name: | signal transducer and activator of transcription 5A |
Gene Aliases: |
AA959963, STAT5 |
Location: |
Chr.11:100859180-100885169 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat5a | NM_001164062.1 | NP_001157534.1 | ||
| NM_011488.3 | NP_035618.1 | |||
| AK134012.1 | BAE21979.1 | |||
| AK154070.1 | BAE32355.1 | |||
| AY299491.1 | AAQ75420.1 | |||
| AY299492.1 | AAQ75421.1 | |||
| BC008998.1 | AAH08998.1 | |||
| BC013274.1 | AAH13274.1 | |||
| EU249543.1 | ||||
| U21103.1 | AAA80590.1 | |||
| U36502.1 | AAA78945.1 | |||
| Z48538.1 | CAA88419.1 |
More Information
Set Membership: |
|

Genomic Map