Assay Details
Target Gene Details
Entrez Gene ID: | 77097 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 |
Gene Aliases: |
3526402J09Rik, 5730590C14Rik |
Location: |
Chr.11:105589986-105929304 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tanc2 | NM_181071.3 | 15 | 2810 | NP_851416.2 |
| AK148972.1 | 2 | 1082 | BAE28706.1 | |
| AK173111.1 | 6 | 1280 | BAD32389.1 | |
| BC089352.1 | 4 | 801 | AAH89352.1 |

Genomic Map