Assay Details
Target Gene Details
Entrez Gene ID: | 21452 |
Gene Name: | transcobalamin 2 |
Gene Aliases: |
AW208754, Tcn-2 |
Location: |
Chr.11:3917077-3932078 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcn2 | NM_001130458.1 | NP_001123930.1 | ||
| NM_001130459.1 | NP_001123931.1 | |||
| NM_015749.3 | NP_056564.1 | |||
| AF090686.1 | AAC61868.1 | |||
| AK133707.1 | BAE21792.1 | |||
| AK143761.1 | BAE25528.1 | |||
| AK147176.1 | BAE27739.1 | |||
| AK161618.1 | BAE36496.1 | |||
| AK170401.1 | BAE41770.1 | |||
| BC003720.1 | AAH03720.1 |

Genomic Map