Assay Details
Target Gene Details
Entrez Gene ID: | 21452 |
Gene Name: | transcobalamin 2 |
Gene Aliases: |
AW208754, Tcn-2 |
Location: |
Chr.11:3917077-3932078 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcn2 | NM_001130458.1 | 8 | 1224 | NP_001123930.1 |
| NM_001130459.1 | 8 | 1193 | NP_001123931.1 | |
| NM_015749.3 | 8 | 1227 | NP_056564.1 | |
| AF090686.1 | 7 | 1154 | AAC61868.1 | |
| AK133707.1 | 8 | 1189 | BAE21792.1 | |
| AK143761.1 | 8 | 1195 | BAE25528.1 | |
| AK147176.1 | 8 | 1198 | BAE27739.1 | |
| AK161618.1 | 8 | 1194 | BAE36496.1 | |
| AK170401.1 | 8 | 1213 | BAE41770.1 | |
| AK185167.1 | 2 | 145 | ||
| AK186678.1 | 2 | 145 | ||
| AK191218.1 | 2 | 242 | ||
| AK198501.1 | 2 | 242 | ||
| AK206141.1 | 2 | 242 | ||
| AK216881.1 | 2 | 242 | ||
| BC003720.1 | 7 | 1100 | AAH03720.1 |

Genomic Map