Assay Details
Target Gene Details
Entrez Gene ID: | 216964 |
Gene Name: | transformation related protein 53 inducible protein 13 |
Gene Aliases: |
2410019G02Rik, Tp53i13 |
Location: |
Chr.11:77508099-77515921 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53i13 | NM_001024920.1 | NP_001020091.1 | ||
| AK213971.1 | ||||
| BC147324.1 | ||||
| BC147325.1 | ||||
| BQ939386.1 |

Genomic Map