Assay Details
Target Gene Details
Entrez Gene ID: | 22415 |
Gene Name: | wingless-type MMTV integration site family, member 3 |
Gene Aliases: |
Int-4, Wnt-3 |
Location: |
Chr.11:103774175-103818021 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wnt3 | NM_009521.2 | 2 | 256 | NP_033547.1 |
| AK043014.1 | 1 | 85 | BAC31433.1 | |
| AK132516.1 | 2 | 259 | ||
| AK141154.1 | 2 | 189 | ||
| AK163878.1 | 1 | 58 | BAE37526.1 | |
| BC141271.1 | 2 | 181 | ||
| BC141274.1 | 2 | 181 | ||
| BC152304.1 | 2 | 192 | ||
| BC152305.1 | 2 | 193 | ||
| BC152537.1 | 2 | 193 | ||
| M32502.1 | 2 | 207 | AAB38109.1 |
More Information
Set Membership: |
|

Genomic Map