Assay Details
Target Gene Details
Entrez Gene ID: | 18789 |
Gene Name: | poly (A) polymerase alpha |
Gene Aliases: |
Pap, PapIII, Plap |
Location: |
Chr.12:105784702-105838949 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Papola | NM_011112.3 | NP_035242.1 | ||
| AK028940.1 | ||||
| AK090096.1 | ||||
| AK132704.1 | BAE21312.1 | |||
| AK134552.1 | ||||
| AK215066.1 | ||||
| U52197.1 | AAC52586.1 |
More Information
Set Membership: |
|

Genomic Map