Assay Details
Target Gene Details
Entrez Gene ID: | 217866 |
Gene Name: | CDC42 binding protein kinase beta |
Gene Aliases: |
MRCKb |
Location: |
Chr.12:111292972-111377718 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42bpb | NM_183016.2 | NP_898837.2 | ||
| AK173103.1 | BAD32381.1 | |||
| AY277589.1 | AAP34402.1 | |||
| BC138035.1 | ||||
| BC138037.1 |
More Information
Set Membership: |
|

Genomic Map