Assay Details
Target Gene Details
Entrez Gene ID: | 13386 |
Gene Name: | delta-like 1 homolog (Drosophila) |
Gene Aliases: |
AW742678, DLK-1, DlkI, FA1, Ly107, Peg9, SCP1, ZOG, pG2, pref-1 |
Location: |
Chr.12:109452823-109463336 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlk1 | NM_001190703.1 | NP_001177632.1 | ||
| NM_001190704.1 | NP_001177633.1 | |||
| NM_001190705.1 | NP_001177634.1 | |||
| NM_010052.5 | NP_034182.2 | |||
| NR_033813.1 | ||||
| AK005522.1 | BAB24096.1 | |||
| AK133664.1 | BAE21772.1 | |||
| AK162458.1 | BAE36929.1 | |||
| AK164935.1 | BAE37973.1 | |||
| AY644392.1 | AAT46420.1 | |||
| BC052159.1 | AAH52159.1 | |||
| CV562715.1 | ||||
| EU434914.1 | ||||
| EU434915.1 | ||||
| EU434916.1 | ||||
| EU434917.1 | ||||
| L12721.1 | AAA37175.1 | |||
| U15980.1 | AAB60495.1 | |||
| Z12171.1 | CAA78162.1 |
More Information
Set Membership: |
|

Genomic Map