Assay Details
Target Gene Details
Entrez Gene ID: | 20403 |
Gene Name: | intersectin 2 |
Gene Aliases: |
AI327390, Ese2, Sh3d1B, Sh3p18, mKIAA1256 |
Location: |
Chr.12:4592638-4713952 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn2 | NM_001198968.2 | NP_001185897.1 | ||
| NM_001198969.2 | NP_001185898.1 | |||
| NM_011365.4 | NP_035495.2 | |||
| AF132479.1 | AAD19747.1 | |||
| AF132480.1 | AAD19748.1 | |||
| AK029325.1 | BAE43269.1 | |||
| AK031390.1 | BAC27381.1 | |||
| AK034367.1 | ||||
| AK122480.1 | BAC65762.3 | |||
| AK161743.1 | ||||
| AK163807.1 | BAE37499.1 | |||
| BC138262.1 | ||||
| BC171950.1 | ||||
| BI872589.1 |
More Information
Set Membership: |
|

Genomic Map