Assay Details
Target Gene Details
Entrez Gene ID: | 20403 |
Gene Name: | intersectin 2 |
Gene Aliases: |
AI327390, Ese2, Sh3d1B, Sh3p18, mKIAA1256 |
Location: |
Chr.12:4592638-4713952 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn2 | NM_001198968.2 | 20 | 2530 | NP_001185897.1 |
| NM_011365.4 | 19 | 2449 | NP_035495.2 | |
| AF132479.1 | 18 | 2189 | AAD19747.1 | |
| AF132480.1 | 18 | 2189 | AAD19748.1 | |
| AK029325.1 | 19 | 2396 | BAE43269.1 | |
| AK050865.1 | 1 | 684 | ||
| AK122480.1 | 14 | 1836 | BAC65762.3 | |
| AK161743.1 | 8 | 867 | ||
| BC138262.1 | 20 | 2507 | ||
| BC171950.1 | 20 | 2507 | ||
| BI872589.1 | 4 | 417 |
More Information
Set Membership: |
|

Genomic Map