Assay Details
Target Gene Details
Entrez Gene ID: | 20403 |
Gene Name: | intersectin 2 |
Gene Aliases: |
AI327390, Ese2, Sh3d1B, Sh3p18, mKIAA1256 |
Location: |
Chr.12:4592638-4713952 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn2 | NM_001198968.2 | 23 | 2981 | NP_001185897.1 |
| NM_011365.4 | 22 | 2900 | NP_035495.2 | |
| AF132479.1 | 21 | 2640 | AAD19747.1 | |
| AF132480.1 | 21 | 2640 | AAD19748.1 | |
| AK122480.1 | 17 | 2287 | BAC65762.3 | |
| AK161743.1 | 11 | 1318 | ||
| BC138262.1 | 23 | 2958 | ||
| BC171950.1 | 23 | 2958 | ||
| BI872589.1 |
More Information
Set Membership: |
|

Genomic Map