Assay Details
Target Gene Details
Entrez Gene ID: | 319670 |
Gene Name: | echinoderm microtubule associated protein like 5 |
Gene Aliases: |
BC027154, C130068M19Rik |
Location: |
Chr.12:98786604-98901494 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eml5 | NM_001081191.1 | NP_001074660.1 | ||
| AK044024.1 | BAC31743.1 | |||
| BC027154.1 | AAH27154.1 | |||
| BC158137.1 |

Genomic Map