Assay Details
Target Gene Details
Entrez Gene ID: | 74855 |
Gene Name: | family with sequence similarity 228, member A |
Gene Aliases: |
4930417G10Rik |
Location: |
Chr.12:4697272-4738468 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam228a |
Target Gene Details
Entrez Gene ID: | 20403 |
Gene Name: | intersectin 2 |
Gene Aliases: |
AI327390, Ese2, Sh3d1B, Sh3p18, mKIAA1256 |
Location: |
Chr.12:4592638-4713952 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 36 - Intron 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn2 | NM_001198968.2 | NP_001185897.1 | ||
| NM_011365.4 | NP_035495.2 | |||
| AF132479.1 | AAD19747.1 | |||
| AF132480.1 | AAD19748.1 | |||
| AK122480.1 | BAC65762.3 | |||
| BC138262.1 | ||||
| BC171950.1 | ||||
| BE650375.1 |
More Information
Set Membership: |
|

Genomic Map