Assay Details
Target Gene Details
Entrez Gene ID: | 65256 |
Gene Name: | ankyrin repeat and SOCS box-containing 2 |
Gene Aliases: |
1110008E15Rik |
Location: |
Chr.12:103321142-103356001 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb2 | NM_023049.1 | 3 | 653 | NP_075536.1 |
| AB221595.1 | 2 | 263 | ||
| AF155353.1 | 1 | 120 | AAD38809.2 | |
| AK083400.1 | 1 | 179 | ||
| BC031161.1 | 3 | 653 | AAH31161.1 |

Genomic Map