Assay Details
Target Gene Details
Entrez Gene ID: | 319670 |
Gene Name: | echinoderm microtubule associated protein like 5 |
Gene Aliases: |
BC027154, C130068M19Rik |
Location: |
Chr.12:98786604-98901494 on Build GRCm38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eml5 | NM_001081191.1 | 41 | 6050 | NP_001074660.1 |
| AK044024.1 | 5 | 703 | BAC31743.1 | |
| BC027154.1 | 4 | 595 | AAH27154.1 | |
| BC158137.1 | 39 | 5522 |

Genomic Map