Assay Details
Target Gene Details
Entrez Gene ID: | 105005 |
Gene Name: | family with sequence similarity 84, member A |
Gene Aliases: |
2310003N02Rik, 4731402F03Rik, AW125753, Nse1 |
Location: |
Chr.12:14147598-14152038 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam84a | NM_029007.2 | 1 | 108 | NP_083283.2 |
| AK014625.1 | 1 | 124 | BAB29470.1 | |
| BC005488.1 | 1 | 108 | AAH05488.1 |

Genomic Map