Assay Details
Target Gene Details
Entrez Gene ID: | 20403 |
Gene Name: | intersectin 2 |
Gene Aliases: |
AI327390, Ese2, Sh3d1B, Sh3p18, mKIAA1256 |
Location: |
Chr.12:4592638-4713952 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn2 | NM_001198968.2 | 7 | 860 | NP_001185897.1 |
| NM_001198969.2 | 7 | 860 | NP_001185898.1 | |
| NM_011365.4 | 7 | 860 | NP_035495.2 | |
| AF132479.1 | 6 | 600 | AAD19747.1 | |
| AF132480.1 | 6 | 600 | AAD19748.1 | |
| AK029325.1 | 7 | 807 | BAE43269.1 | |
| AK034367.1 | 7 | 861 | ||
| AK042449.1 | 7 | 830 | BAC31264.1 | |
| AK122480.1 | 2 | 247 | BAC65762.3 | |
| AK163807.1 | 7 | 834 | BAE37499.1 | |
| BC138262.1 | 7 | 837 | ||
| BC171950.1 | 7 | 837 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map