Assay Details
Target Gene Details
Entrez Gene ID: | 12499 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 5 |
Gene Aliases: |
AI196558, AI987697, Cd39l4, ER-UDPase, NTPDase-5, NTPDase5, Pcph, mNTPase |
Location: |
Chr.12:84373875-84409029 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Entpd5 | NM_001026214.2 | NP_001021385.1 | ||
| NM_001286049.1 | NP_001272978.1 | |||
| NM_001286058.1 | NP_001272987.1 | |||
| NM_007647.3 | NP_031673.2 | |||
| AF006482.1 | AAC05181.1 | |||
| AF136571.1 | AAK82949.1 | |||
| AJ238636.1 | CAB45533.1 | |||
| AK002618.1 | BAB22234.1 | |||
| AK031581.1 | BAC27461.1 | |||
| AK036641.1 | BAC29515.1 | |||
| AK037736.1 | BAC29861.1 | |||
| AK045828.1 | BAC32507.1 | |||
| AK079267.1 | BAC37592.1 | |||
| AK080265.1 | BAC37862.1 | |||
| AK081435.1 | BAC38219.1 | |||
| AK088455.1 | BAC40362.1 | |||
| AK160950.1 | BAE36110.1 | |||
| AK163688.1 | BAE37457.1 | |||
| BC015247.1 | AAH15247.1 |
More Information
Set Membership: |
|

Genomic Map